Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
Brand: Macherey-Nagel™ 726038.25
This item is not returnable.
View return policy
Specifications
0.4 mm | |
0.25 mm | |
G1, G2, G38 | |
340°C to 360°C | |
25 m |
GC Capillary Column | |
Optima™ 1 | |
Separation of Components According to Boiling Points Analysis | |
0.1 μm | |
1 Each |
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ 1 Capillary Column for GC > I.D.: 0.25mm; Length: 25m; Film Thickness: 0.1um
Spot an opportunity for improvement?Share a Content Correction