Learn More
Thermo Scientific™ Orion™ Pro Star Series Meter Accessories
Select Thermo Scientific™ Orion™ Pro Star series meter accessories to support your Orion Pro Star PH211, EC212, DO213, PI214, PC215, PD216, PI217 and MP218 bench meters.
Brand:  Thermo Scientific™ PSTAR-PWR
 View more versions of this product
Product Code. 30245797
  • 48.00 EUR / 1 set
Explore more special offers
Description
Show More
Specifications
Specifications
Power Supply
50/60 Hz
100/115/220/230 V
Orion Pro Star series bench electrochemistry meters
Replace your power supply as needed
1 meter power supply with US/Japan, Euro, UK/Singapore, Australia/New Zealand and China wall plug plates
US/Japan, Euro, UK/Singapore, Australia/New Zealand and China wall plugs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Pro Star Series Meter Accessories > Pro Star universal power supply