Learn More
Osmolality Standard, 200mOsm/kg H2O, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
12 x 5 mL
Specifications
Specifications
Packaging | Ampoules |
Quantity | 12 x 5 mL |
Physical Form | Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
OSM20024H1 | Certificate of Analysis | 04/03/2025 | RE-OSM-200 |
OSM20023M1 | Certificate of Analysis | 29/08/2024 | RE-OSM-200 |
OSM20022K1 | Certificate of Analysis | 09/01/2024 | RE-OSM-200 |
OSM20022B1 | Certificate of Analysis | 30/04/2022 | RE-OSM-200 |
OSM20021E1 | Certificate of Analysis | 29/03/2022 | RE-OSM-200 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Product Content Correction
Product Title
Osmolality Standard, 200mOsm/kg H2O, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction