missing translation for 'onlineSavingsMsg'
Learn More
Oxalic acid dihydrate, ExpertQ, Scharlab Product Code.: 11358624
compliance-icons compliance-icons

Oxalic acid dihydrate, ExpertQ, Scharlab

Product Code. 11358624
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11358624

Brand: Scharlab AC17201000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES O.O.OC(=O)C(O)=O
IUPAC Name oxalic acid dihydrate
Molecular Weight (g/mol) 126.06
Grade ExpertQ
Quantity 1 kg
compliance-icons compliance-icons
Product Identifier
  • Oxalic acid dihydrate, reagent grade, ACS,ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H318-Causes serious eye damage.
  • H302 + H312-Harmful if swallowed or in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxalic acid dihydrate, ExpertQ, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.