Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sachets
Oxoid AnaeroGen Compact Sachet is a simple anaerobic gas generator system for the anaerobic incubation of up to 4 petri dishes or an identification panel.
Brand: Thermo Scientific™ AN0020C
66.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Oxoid AnaeroGen Compact Sachet is a simple anaerobic gas generator system for the anaerobic incubation of up to 4 petri dishes or an identification panel.
- Use 1 sachet / Compact Plastic Pouch
- Use with Compact Plastic Pouch, Part No. AG0020C and AnaeroGen™ Compact Sealing Clip, Part No. AN0005C
Includes: 20 sachets.
Specifications
AnaeroGen Compact Sachets | |
20 Sachets | |
10 Each/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6223635 | Certificate of Analysis | 31/01/2025 | AN0020C |