Learn More
Thermo Scientific™ Oxoid™ AnaeroJar™ O-Ring
Oxoid AnaeroJar O-Ring is for use with AnaeroJar 2.5L, Part No. AG0025A.
Brand:  Thermo Scientific™ AG0030A
42.69 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10118052
  • 45.45 EUR / Pack of 5
Estimated Shipment: 13-05-2025
Description
Oxoid AnaeroJar O-Ring is for use with AnaeroJar 2.5L, Part No. AG0025A.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AnaeroJar O-ring
Thermo Scientific Oxoid
5/Pk.
SDS
Documents Certificates
Product Title
Thermo Scientific™ Oxoid™ AnaeroJar™ O-Ring > 5/Pk.