Learn More
Parimifasor, MedChemExpress
MedChemExpress Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.
Brand: Medchem Express HY-109098
Description
Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.Specifications
Parimifasor | |
White | |
C18H11Cl2F4N5O | |
LYC30937 | |
O=C(C1=CC(Cl)=CC=C1)N/C(NC2=NNC(C(F)(F)F)=C2)=N/C3=CC(Cl)=CC(F)=C3 | |
460.21 | |
Research | |
COVID-19-immunoregulation |
1796641-10-5 | |
Research | |
100 mg | |
DMSO : 66.67 mg/mL (144.87 mM; Need ultrasonic) | |
460.21 | |
98.07% | |
Solid |