Learn More
Brand™ PCR Plate Combo Pack PROMO
Provides additional savings than when plates and fiilm are purchased individually. Brand™ PCR Plate Combo Pack PROMO includes 96-well PCR plates available in different skirt and profile types along with high quality qPCR sealing film.
Brand:  Brand™ 781231
 View more versions of this product
Product Code. 15504419
  • 275.00 EUR / 1 set
Explore more special offers
Includes: 96-well PCR plates, PCR-sealing film
Description
Description
Premium PCR/qPCR transparent plates
  • 96-well
  • Skirted, non-skirted, or semi-skirted
  • Low, standard or elevated rim profile
  • Ultra-thin walled for quick and constant thermal transfer
  • Free of DNase, DNA, RNase and endotoxins
  • No warping, no wiggling, suitable for automation
Includes qPCR PCR sealing film:
  • Highly transparent for qPCR
  • Press to seal
  • Convenient end tabs
  • No annoying adhesive residues
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Non-skirted PCR plates
96
PCR Combo Pack
50/Pk.
96-well