Learn More
Brand™ PCR Plate Combo Pack PROMO
Provides additional savings than when plates and fiilm are purchased individually. Brand™ PCR Plate Combo Pack PROMO includes 96-well PCR plates available in different skirt and profile types along with high quality qPCR sealing film.
275.00 EUR
Specifications
Quantity 50/Pk.
No. of Wells 96
Format 96-well
Product Type PCR Combo Pack
View More Specs
Includes: 96-well PCR plates, PCR-sealing film
Products 5
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15504419
View Documents
Brand™
781231
Non-skirted PCR plates
275.00 EUR
1 set
15514419
View Documents
Brand™
781232
Elevated rim low profile plates
275.00 EUR
1 set
15524419
View Documents
Brand™
781233
Semi-Skirted, Elevated, Low profile plates
275.00 EUR
1 set
15534419
View Documents
Brand™
781234
Standard profile plates
275.00 EUR
1 set
15594409
View Documents
Brand™
781230
Non-Skirted, Low profile plates
275.00 EUR
1 set
Description
Description
Premium PCR/qPCR transparent plates
  • 96-well
  • Skirted, non-skirted, or semi-skirted
  • Low, standard or elevated rim profile
  • Ultra-thin walled for quick and constant thermal transfer
  • Free of DNase, DNA, RNase and endotoxins
  • No warping, no wiggling, suitable for automation
Includes qPCR PCR sealing film:
  • Highly transparent for qPCR
  • Press to seal
  • Convenient end tabs
  • No annoying adhesive residues
Specifications
Specifications
50/Pk.
96
96-well
PCR Combo Pack