Learn More
Brand™ PCR Plate Combo Pack PROMO
Provides additional savings than when plates and fiilm are purchased individually. Brand™ PCR Plate Combo Pack PROMO includes 96-well PCR plates available in different skirt and profile types along with high quality qPCR sealing film.
Brand: Brand™ 781232
Includes: 96-well PCR plates, PCR-sealing film
Description
Premium PCR/qPCR transparent plates- 96-well
- Skirted, non-skirted, or semi-skirted
- Low, standard or elevated rim profile
- Ultra-thin walled for quick and constant thermal transfer
- Free of DNase, DNA, RNase and endotoxins
- No warping, no wiggling, suitable for automation
- Highly transparent for qPCR
- Press to seal
- Convenient end tabs
- No annoying adhesive residues
Specifications
Elevated rim low profile plates | |
96 | |
PCR Combo Pack |
50/Pk. | |
96-well |