Learn More
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand:  Fisherbrand™ 22 08 0794
33.77 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11797617
  • 36.97 EUR / Pack of 1000
In Stock
Description
  • PE Snap Cap
  • Transparent
  • Closed top
  • Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
Polyethylene
ND22
22 mm
Snap Cap
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    3
    Hours
    13
    Minutes
    24
    Seconds
    35
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
    Days
    78
    Hours
    13
    Minutes
    24
    Seconds
    35
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    34
    Hours
    13
    Minutes
    24
    Seconds
    35
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Days
    3
    Hours
    13
    Minutes
    24
    Seconds
    35
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial > 22mm