Learn More
Cytiva PEEK Finger Tight Connector
Use With Tubing o.d. 1/16 in. Cytiva PEEK Finger Tight Connector provides the tightest connections.
Brand: Cytiva 18-1112-55
113.05 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
- Packsize 10 Connections
- Fingertight 1/16 in.
- Polyetheretherketone (PEEK)
Specifications
1/16 in. Male |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
12Hours
16Minutes
48Seconds
0 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
42Hours
16Minutes
48Seconds
0