Learn More
3M™ PELTOR™ Rechargeable NimH Battery Pack ACK053
3M™ PELTOR™ Rechargeable NimH Battery Pack ACK053 designed for use with 3M™ PELTOR™ WS Headset XP, WS ProTac XP, WS5 Adapter, WS LiteCom headsets.
Brand: 3M™ 7100075338
54.79 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Rechargeable NiMH battery pack, 2500 mAh
- For use with 3M™ PELTOR™ 3M™ PELTOR™ WS Headset XP, WS ProTac XP, WS5 Adapter, WS LiteCom Headsets
- PELTOR™ FR05 charger supplied separately
- 1900mAH
- Charging time 5-6 hours
- NiMh Battery
Specifications
Battery Pack | |
3M PELTOR WS Headset XP, WS ProTac XP, WS5 Adapter, WS LiteCom Headsets |