Learn More
PEMT 293T Cell Overexpression Lysate (Denatured), Abnova
Brand:  Abnova H00010400-T01.100uL
Product Code. 16175012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Western Blotting
Specifications
Specifications
293T
PEMT 293T Cell Transient Overexpression Lysate(Denatured)
26.07
pCMV-PEMT full-length
100 μL
Human
Western Blot
BC000557
6.8
Transient overexpression cell Lysate was tested with Anti-PEMT antibody (H00010400-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
PEMT 293T Cell Overexpression Lysate (Denatured), Abnova > 100μL