Learn More
Sampling Systems™ Pendulum Beaker
These pendulum beakers automatically swing to the horizontal position, meaning that your water samples are less likely to spill when using the TeleScoop.
Brand: Sampling Systems™ 5065C-502
This item is not returnable.
View return policy
Description
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Specifications
Swabbing Template | |
Autoclavable | |
Reusable | |
Non-sterile |
Polypropylene | |
500 mL | |
No |
Product Content Correction
Product Title
Sampling Systems™ Pendulum Beaker > Pendulum Beaker; Capacity (Metric): 500 mL; Graduated: No; Material: Polypropylene
Spot an opportunity for improvement?Share a Content Correction