missing translation for 'onlineSavingsMsg'
Learn More
Honeywell Safety Products™ Perfect Poly™ Finger Gloves Product Code.: 11881741
Estimated Shipment: 09-06-2025
to see stock.
Add to basket

Honeywell Safety Products™ Perfect Poly™ Finger Gloves

Product Code. 11881741
7
Click to view available options
Size:
6
7
8
This item is not returnable. View return policy

Product Code. 11881741

Brand: Honeywell Safety Products™ 223224007

Estimated Shipment: 09-06-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

White polyamide knitted gloves with polyurethane-coated fingertips provide dexterity, a snug fit, and good grip for precision work. Honeywell Safety Products™ Perfect Poly™ Finger Gloves are knitted for breathability and comfort and offer excellent abrasion and tear resistance.

  • White, non-pilling polyamide knitted glove suitable for precision work.
  • Suitable for handling fine products in a dry environment.
  • Excellent abrasion and tear resistance.
  • Silicone-free and suitable for painting environments.
  • White polyurethane coating on fingertips for good grip.
  • Tight knit provides comfort, support, dexterity, touch and limits dirt penetration.
  • Ventilated back and palm optimize breathability.
  • Elastic wrist.
  • White color enhances the glove's hygiene and cleanliness.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY I Minor risks, EN 420-2003 Basic standard, EC Category PPE 1, EC Certificate Number Autocertification
Classification Dexterity: 5
Coated Area Fingers
Description Gauge 15
Coating Material Polyurethane
Color White
Material Polyurethane, Polyamide
Product Type Gloves
Size 7
For Use With (Application) Automotive, Aviation and Utility industries
Includes Elasticized wrist
Quantity 10Pair/PK
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell Safety Products™ Perfect Poly™ Finger Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.