missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluorotributylamine, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 311-89-7 |
Molecular Formula | C12F27N |
Quantity | 10 g |
Synonym | perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine |
InChI Key | RVZRBWKZFJCCIB-UHFFFAOYSA-N |
SMILES | C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
IUPAC Name | 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine |
Molecular Weight (g/mol) | 671.096 |
PubChem CID | 9397 |
ChEBI | CHEBI:38854 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Perfluorotributylamine, MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction