Learn More
Cole-Parmer™ PFA Threaded Reducing Bushing
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-10
 View more versions of this product
Product Code. 11858073
  • 147.00 EUR / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
NPT(M) x NPT(F) Connection
PFA
3/4 in.
3/4 in. NPTM x 1/2 in. NPTF
Threaded Reducing Bushing
Autoclavable
1/2 in.
Autoclave or Ethylene Oxide
Translucent