missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKFB1/PFKFB4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5104524
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PFKFB1/4.
This gene encodes a member of the family of bifunctional 6-phosphofructo-2-kinase:fructose-2,6-biphosphatase enzymes. The enzyme forms a homodimer that catalyzes both the synthesis and degradation of fructose-2,6-biphosphate using independent catalytic domains. Fructose-2,6-biphosphate is an activator of the glycolysis pathway and an inhibitor of the gluconeogenesis pathway. Consequently, regulating fructose-2,6-biphosphate levels through the activity of this enzyme is thought to regulate glucose homeostasis. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
PFKFB1/PFKFB4 | |
Polyclonal | |
Unconjugated | |
Pfkfb1 | |
6PF-2-K/Fru-2,6-P2ase 1; 6PF-2-K/Fru-2,6-P2ase 4; 6PF-2-K/Fru-2,6-P2ase liver isozyme; 6PF-2-K/Fru-2,6-P2ase testis-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4; bifunctional enzyme with kinase and biphosphatase activities; C230090D14; F6PK; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase:fructose-2,6-bisphosphatase; HL2K; PFK/FBPase 1; PFK/FBPase 4; PFK-2/FBPase-2 gene A; Pfkfb01; Pfkfb1; Pfkfb4; PFRX | |
Rabbit | |
Affinity chromatography | |
RUO | |
18639, 24638, 270198, 5207, 5210, 54283 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P07953, P16118, P25114, P70266, Q16877, Q6DTY7 | |
Pfkfb1, Pfkfb4 | |
A synthesized peptide derived from human PFKFB1(Accession P16118), corresponding to amino acid residues I329-I379. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PFKFB1/PFKFB4 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction