Learn More
Schmidlin™ PG Plus FID Station H2 and Zero Air Generator
This dual gas generator produces a continuous supply of high purity hydrogen and zero air.
Brand:  Schmidlin™ DB-FHG250-EU
Encompass_Preferred
 View more versions of this product
Product Code. 17758879
  • 14388.54 EUR / Each
Explore more special offers
Description
Description
  • Increase efficiency and improve performance
  • On-demand supply 24/7
  • H2 flow rate and zero air up to 5 L/min.
  • THC Pressure: 11 barg (160 psig)
  • Permeation membrane drying system
  • USB connectivity
  • Easy to install with 'plug and play' functionality
  • Ideal for all GC detector applications
Specifications
Specifications
68 cm
15 cm
11 bar
69 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
115 W
250 mL/min.
35°C
27 in.
Outlet: 1/8 in.
>99.9996%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Schmidlin™ PG Plus FID Station H2 and Zero Air Generator