Learn More
Christeyns™ Phago'Gel SPS
Phago'Gel SPS is intended for the hygienic surgical disinfection of hands by friction as an alternative to simple or hygienic hand washing, in the absence of water, during serial contact, or before any aseptic gesture.
Brand: Christeyns™ P60673
View more versions of this product
This item is not returnable.
View return policy
Description
- High tolerance
- Fast to use
- Bactericidal, fungicidal, mycobactericidal and virucidal
- Fragrance-free, dye-free, phenoxyethanol-free and paraben-free
Specifications
Alcohol-based Hand Sanitizer Gel | |
Gel | |
300 mL |
Colorless | |
20 Bottles with 3 mL Pump |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
SDS_17800255_EN.pdf
File size: 246 kB
Product Content Correction
Product Title
Christeyns™ Phago'Gel SPS > Phago'Gel SPS; Volume (Metric): 300 mL; Quantity: 20 Bottles with 3 mL Pump
Spot an opportunity for improvement?Share a Content Correction