Learn More
Christeyns™ Phago'Gel SPS
Phago'Gel SPS is intended for the hygienic surgical disinfection of hands by friction as an alternative to simple or hygienic hand washing, in the absence of water, during serial contact, or before any aseptic gesture.
Brand: Christeyns™ C65054
View more versions of this product
This item is not returnable.
View return policy
Description
- High tolerance
- Fast to use
- Bactericidal, fungicidal, mycobactericidal and virucidal
- Fragrance-free, dye-free, phenoxyethanol-free and paraben-free
Specifications
Hand Disinfectant Gel | |
Gel | |
5 L |
Colorless | |
2 Cans |
SDS_17840255_EN.pdf
File size: 246 kB
Product Content Correction
Product Title
Christeyns™ Phago'Gel SPS > Phago'Gel SPS; Volume (Metric): 5 L; Quantity: 2 Cans
Spot an opportunity for improvement?Share a Content Correction