missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
Brand: MP Biomedicals 0215612810
This item is not returnable.
View return policy
Description
Specifications
>99% | |
100929-99-5 | |
Store at 0°C. | |
519.471 | |
10mg |
0.854 | |
White | |
Powder | |
Phe-Arg-β-naphthylamide dihydrochloride |