missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
44.20 EUR - 481.24 EUR
Specifications
Purity | >99% |
---|---|
Assay | 0.854 |
CAS | 100929-99-5 |
Color | White |
Content And Storage | Store at 0°C. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433492
|
MP Biomedicals
0215612810 |
10mg |
44.20 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443492
|
MP Biomedicals
0215612825 |
25mg |
76.85 EUR
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453492
|
MP Biomedicals
0215612850 |
50mg |
145.22 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463492
|
MP Biomedicals
0215612883 |
250mg |
481.24 EUR
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
>99% | |
100929-99-5 | |
Store at 0°C. | |
519.471 |
0.854 | |
White | |
Powder | |
Phe-Arg-β-naphthylamide dihydrochloride |