Learn More
Corning™ Phenolic Screw Threaded Caps
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Brand: Corning™ 9999-132
This item is not returnable.
View return policy
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
13-415 | |
Screw Cap | |
Phenolic | |
13.5mm | |
36 Pack | |
36 Pack |
Rubber | |
Autoclavable | |
Black | |
36 Pack | |
36 Pack | |
Culture Tubes |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Corning™ Phenolic Screw Threaded Caps > Height: 13.5mm
Spot an opportunity for improvement?Share a Content Correction