Learn More
Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4
Supplied as exactly pre-weighed tablets with a final buffer volume of 500 or 1000ml. Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4 is dissolved in deionized water to yield a solution of sodium chloride, potassium chloride, Tween™ 20 and phosphate buffer.
Brand:  Medicago 09-9410-100
 View more versions of this product
Product Code. 11300439
  • 341.00 EUR / Pack of 100
Edge
Explore more special offers
Description
Description
PBS-T buffer contains the non-ionic detergent Tween™ 20 which acts through blocking. It has the ability to reduce non-specific binding and protein-protein interaction during the wash step in protein and immunoassay procedures such as ELISA and Western blotting. Decreasing the non-specific binding and staining makes ELISA results and blots easier to interpret.
  • Formulated from analytical grade reagents
  • PBS with non-ionic detergent
  • Specifically developed for immunoassays
  • Exactly pre-weighed tablets
  • Dissolve and go for greater convenience
Specifications
Specifications
100 x 1000 mL Tablets
ELISA, Western Blot, Plate-based Assays, Protein-plate Coating
7.4 at 25°C
Tablets
NaCl (0.14M), KCl (0.0027M), PO4 (0.01M), Tween™ 20 (0.05%)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4 >