Learn More
PI4KIIIbeta-IN-10, MedChemExpress
MedChemExpress PI4KIIIbeta-IN-10 is a potent PI4KIIIβ inhibitor with an IC50 of 3.6 nM.
Brand: Medchem Express HY-100198
Description
PI4KIIIbeta-IN-10 is a potent PI4KIIIβ inhibitor with an IC50 of 3.6 nM.Specifications
PI4KIIIbeta-IN-10 | |
White | |
C22H25N3O5S2 | |
DMSO : ≥ 30 mg/mL (63.08 mM) | |
475.58 | |
98.04% | |
Powder |
1881233-39-1 | |
Research | |
5 mg | |
COC1=CC=C(C2=C(C)N=C(NC(C(C)(C)C)=O)S2)C=C1S(NC3=CC=C(O)C=C3)(=O)=O | |
475.58 | |
Research | |
COVID-19-immunoregulation |