Learn More
PI4KIIIbeta-IN-10, MedChemExpress
MedChemExpress PI4KIIIbeta-IN-10 is a potent PI4KIIIβ inhibitor with an IC50 of 3.6 nM.
Brand:  Medchem Express HY-100198
Encompass_Preferred
 View more versions of this product
Product Code. 16405587
  • 473.85 EUR / 5mg
Description
PI4KIIIbeta-IN-10 is a potent PI4KIIIβ inhibitor with an IC50 of 3.6 nM.
Specifications
Specifications
PI4KIIIbeta-IN-10
White
C22H25N3O5S2
DMSO : ≥ 30 mg/mL (63.08 mM)
475.58
98.04%
Powder
1881233-39-1
Research
5 mg
COC1=CC=C(C2=C(C)N=C(NC(C(C)(C)C)=O)S2)C=C1S(NC3=CC=C(O)C=C3)(=O)=O
475.58
Research
COVID-19-immunoregulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PI4KIIIbeta-IN-10, MedChemExpress