Learn More
Thermo Scientific™ Pierce™ Alkylating Reagents
Alkylating reagents provided in single-use, pre-weighed formats to streamline protein preparation workflows.
Brand: Thermo Scientific™ A39270
72.00 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
View more versions of this product
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific Pierce Iodoacetamide or Chloroacetamide are provided in a no-weigh format, where small amounts of reagent are pre-weighed and packaged for single use. This single-use format prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. The format enables use of a fresh vial of reagent each time, and eliminates the hassle of weighing out small amounts of reagent.
Alkylating reagents iodoacetamide and chloracetamide are sulfhydryl-reactive alkylating reagents used to block reduced cysteine residues for protein characterization and peptide mapping. Alkylation with iodoacetamide or chloracetamide after cysteine reduction results in the covalent addition of a carbamidomethyl group (57.07 Da) and prevents the formation of disulfide bonds. Reducing agents added after alkylation will react with excess iodoacetamide.
Specifications
Alkylating Reagent | |
10 x 2 mg | |
Chloroacetamide | |
Pierce™ |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZK393139 | Certificate of Analysis | 16/12/2024 | A39270 |
ZH393138 | Certificate of Analysis | 27/08/2024 | A39270 |
ZD390122 | Certificate of Analysis | 17/05/2024 | A39270 |
ZC391285 | Certificate of Analysis | 21/03/2024 | A39270 |
ZB386708 | Certificate of Analysis | 22/02/2024 | A39270 |