Learn More
Piperaquine tetraphosphate tetrahydrate, MedChemExpress
MedChemExpress Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.
Brand:  Medchem Express HY-B1896B
Encompass_Preferred
 View more versions of this product
Product Code. 16424637
  • 187.65 EUR / 50mg
Description
Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.
Specifications
Specifications
Piperaquine tetraphosphate tetrahydrate
915967-82-7
50 mg
C29H32Cl2N6.4H3O4P.4H2O
H2O : 6 mg/mL (6.00 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (5.00 mM; Need ultrasonic and warming)
999.6
98.0%
Solid
White
Research
Neuroscience-Neuromodulation
ClC1=CC=C2C(N3CCN(CCCN4CCN(C5=CC=NC6=CC(Cl)=CC=C56)CC4)CC3)=CC=NC2=C1.OP(O)(O)=O.[4].[4 H2O]
999.6
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Piperaquine tetraphosphate tetrahydrate, MedChemExpress