Learn More
Piperaquine tetraphosphate tetrahydrate, MedChemExpress
MedChemExpress Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.
Brand: Medchem Express HY-B1896B
Description
Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.Specifications
Piperaquine tetraphosphate tetrahydrate | |
915967-82-7 | |
50 mg | |
C29H32Cl2N6.4H3O4P.4H2O | |
H2O : 6 mg/mL (6.00 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (5.00 mM; Need ultrasonic and warming) | |
999.6 | |
98.0% |
Solid | |
White | |
Research | |
Neuroscience-Neuromodulation | |
ClC1=CC=C2C(N3CCN(CCCN4CCN(C5=CC=NC6=CC(Cl)=CC=C56)CC4)CC3)=CC=NC2=C1.OP(O)(O)=O.[4].[4 H2O] | |
999.6 | |
Research |
Product Suggestions
Customers who viewed this item also viewed.