Learn More
Sartorius™ Pipette Calibration Kit
For use with balances with a readability of 0.01 mg.
Brand: Sartorius™ YCP06SQ
1205.63 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory item
- Kit consists of a moisture trap and all required adapters
Specifications
Lab Balance with 0.01 mg readability | |
Weighing | |
Moisture trap and adapters | |
Pipette Calibration Kit |
Product Content Correction
Product Title
Sartorius™ Pipette Calibration Kit > For readability of 0.01 mg
Spot an opportunity for improvement?Share a Content Correction