Learn More
BRAND™ Pipette Tray
Drawer organizer
Brand: BRAND™ 768515
65.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
PVC | |
355 mm |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Product Content Correction
Product Title
BRAND™ Pipette Tray > Dimensions (L x W): 355 x 300 mm
Spot an opportunity for improvement?Share a Content Correction