Learn More
Cole-Parmer™ Plas-Labs™ 800-PRV Glove Box Pressure Relief Valve
Useful for eliminating unwanted positive pressure build-up in glove boxes
Brand: Cole-Parmer™ 34749-68
This item is not returnable.
View return policy
Description
- Acrylic relief valve.
- Features 1/8in. NPT stainless steel inlet and outlet ports.
- Exhausted air can be directed away from operators by a hose connection to the outlet port.
Specifications
NPT(M) x NPT(F) | |
1 in. | |
1/8 in. NPT(F) | |
2 in. |
Stainless Steel | |
1/8 in. NPT(M) | |
1 in. | |
Glove Box Pressure Relief Valve, 1/8 in. NPT Stainless Steel |
Product Content Correction
Product Title
Cole-Parmer™ Plas-Labs™ 800-PRV Glove Box Pressure Relief Valve > 1/8 in.
Spot an opportunity for improvement?Share a Content Correction