Learn More
Microspec™ Plastic Culture Loop
Microseal pouches of sterile plastic culture loops.
Brand: Microspec™ C2G
This item is not returnable.
View return policy
Description
- 12,000 Plastic Culture Loops
- Microseal Pouch of 20
- 200 mm length
- Sterile EtO
Specifications
Single-use | |
7.8 in. | |
Plastic | |
Standard | |
Inoculating Loop | |
Not Autoclavable |
1 μL | |
200 mm | |
Sterile | |
Blue | |
Yes |
Product Content Correction
Product Title
Microspec™ Plastic Culture Loop > Volume: 1 uL
Spot an opportunity for improvement?Share a Content Correction