missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXNB1 Monoclonal Antibody (559830), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524250
266.01 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
PLXNB1 Monoclonal antibody specifically detects PLXNB1 in Human samples. It is validated for Flow Cytometry.
Specifications
PLXNB1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O43157 | |
Plxnb1 | |
NS0 mouse myeloma cell line transfected with human Plexin B1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
559830 | |
Unconjugated | |
Plxnb1 | |
2900002G15Rik; AU042020; Kiaa0407; mKIAA0407; plexin 1; plexin 5; plexin B1; plexin-A1; PLEXIN-B1; PLXN5; Plxnb1; Semaphorin receptor SEP; SEP | |
Mouse | |
Protein A/G | |
RUO | |
5364 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLXNB1 Monoclonal Antibody (559830), Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction