missing translation for 'onlineSavingsMsg'
Learn More

Poly-L-Lysine Hydrobromide [MW 4,000-15,000], MP Biomedicals

88.55 EUR - 926.00 EUR

Chemical Identifiers

CAS 25988-63-0
Molecular Formula C18H38N6O4
Molecular Weight (g/mol) 402.54
InChI Key WBSCNDJQPKSPII-KKUMJFAQSA-N
Synonym lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
PubChem CID 72363
IUPAC Name (2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
SMILES C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478340
View Documents
MP Biomedicals
0210269125
25 mg
88.55 EUR
25mg
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
11488340
View Documents
MP Biomedicals
0210269180
100 mg
277.00 EUR
100mg
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
 
11498340
View Documents
MP Biomedicals
0210269190
500 mg
926.00 EUR
500mg
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Chemical Identifiers

25988-63-0
402.54
lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
(2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
C18H38N6O4
WBSCNDJQPKSPII-KKUMJFAQSA-N
72363
C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N

Specifications

25988-63-0
lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N
402.54
4,000 to 15,000
C18H38N6O4
WBSCNDJQPKSPII-KKUMJFAQSA-N
(2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
72363
Poly-L-Lysine Hydrobromide
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 0°C

SDS
Documents

Documents

Product Certifications