Learn More
Shield Medicare™ Polyester Isolator Cleaning Tool
Brand: Shield Medicare™ 3036230
561.00 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
This item is not returnable.
View return policy
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%
Product Content Correction
Product Title
Shield Medicare™ Polyester Isolator Cleaning Tool > Sterile
Spot an opportunity for improvement?Share a Content Correction