missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene Glycol 400, Synthesis Grade, Scharlab Product Code.: 11348654

Polyethylene Glycol 400, Synthesis Grade, Scharlab

Product Code. 11348654
5 L
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy

Product Code. 11348654

Brand: Scharlab PO0035005P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, chromatography, synthesis of organic products, in the food industry and in cosmetics.

  • Synonyms: Carbowax, PEG, Polyoxyethylene glycol, Polyglycol, Polyethylene oxide
  • Molecular Formula: HO(C2H4O)nH
  • Molar Mass: 380–420g/mol
  • CAS: 25322-68-3
  • EIN ECS No.: 203-473-3
  • Hydroxyl Number: 267–295
  • Residue on Ignition: max. 0.02%

Specifications

CAS 25322-68-3
Melting Point 4°C to 8°C
Density 1.13g/cm3
Boiling Point >250°C
Flash Point 240°C
Ignition Point ∼370°C
Molecular Formula (C2H4O)n
MDL Number MFCD01779601
Quantity 5 L
Synonym Carbowax, PEG, Polyoxyethylene glycol, Polyglycol, Polyethylene oxide
Solubility Information Miscible in water at 20°C
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES [H]OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Formula Weight 380–420g/mol
Vapor Pressure <0.1hPa (at 20°C)
Grade Synthesis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene Glycol 400, Synthesis Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.