Learn More
Macherey-Nagel™ Polygram CEL 300-10 Glass Plates
Brand:  Macherey-Nagel™ 808013
Product Code. 10266761
  • 195.00 EUR / Pack of 25
Estimated Shipment: 13-05-2025
Specifications
25 Pk.
Glass Plate
0.1 mm
20 x 20 cm
Documents
One moment while we fetch your results.
Product Title
Macherey-Nagel™ Polygram CEL 300-10 Glass Plates > Plate Size: 200 x 200mm