missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PP Wide Mouth Erlenmeyer Flask

Mix and store solutions and cell cultures safely. VITLAB™ PP Wide Mouth Erlenmeyer Flask features a wide opening for easy filling and a screw cap for secure storage. Transparent polypropylene flasks with a printed scale make identification simple. Available in a range of sizes.

Brand:  VITLAB™ 669941

 View more versions of this product

Product Code. 10569261

This item is not returnable. View return policy

Description

Description

Erlenmeyer flasks feature a flat bottom, conical body and narrowed neck. These plastic flasks are designed for maximum user convenience, with a wide-mouth opening, screw cap and printed scale. Flasks are constructed of break-resistant, transparent polypropylene and are individually packaged in clearly labeled polyethylene bags.

Wide-mouth top with a screw cap
  • Large opening makes adding liquids and powders easy, even without a funnel
    GL threads support a strong hold on the screw cap for secure storage
  • Can also be used with NS stoppers (not included)
Highly transparent with printed blue scale
  • Easy content identification, measurement and monitoring
  • To preserve markings, cleaning at no higher than 60°C is recommended
Constructed of durable polypropylene
  • Break-resistant plastic is far safer than glass
  • Displays excellent chemical resistance
  • Heat resistant and suitable for use in microwaves ovens
  • Light weight, easy to use and recyclable
Ideal for a range of lab applications
  • Well suited for storage and cultivation of cell cultures
  • Easy mixing of solutions by swirling or shaking without risk of spillage
  • Reduced solvent loss with boiling liquids and recrystallization
  • Suitable for use under a burette during titration
Specifications

Specifications

Polypropylene
Printed graduations
Polypropylene
6
Wide
GL 52; NS 45/40
Transparent
500 mL
100 mL
Erlenmeyer
Screw
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VITLAB™ PP Wide Mouth Erlenmeyer Flask > Capacity: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.