Learn More
Brand™ Polystyrene Inoculation Loop
Permits gentle streaking without damaging the nutrient surface. BRAND™ Polystyrene Inoculation Loops are highly flexible, with loop at one end and needle at the other or loops at both ends.
Brand:  Brand™ 452201
69.16 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 10160721
  • 79.45 EUR / Pack of 1000
Estimated Shipment: 24-04-2025
Description
  • Manufactured from polystyrene.
  • For inoculation of nutrient media.
  • Available either with loop at one end and needle at the other or loops at both ends.
  • High flexibility permits gentle streaking without damaging the nutrient surface.
  • y-ray sterilized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
Polystyrene
White
Needle
1 μL
Sterile
Inoculating Loop
1000/Pk.
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Days
    27
    Hours
    16
    Minutes
    57
    Seconds
    36
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Brand™ Polystyrene Inoculation Loop > Volume: 1μL