Learn More
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
271.00 EUR - 326.00 EUR
Specifications
Product Type | Hygiene Sponge Sampling Kit |
---|---|
Media Type | Neutralizers |
Sterility | Sterile |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17956549
|
LABPLAS
KSS-67910-DE |
None |
271.00 EUR
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17936649
|
LABPLAS
KSS-67915-DE |
Gloves |
326.00 EUR
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Polyurethane Sani-Stick
- Saturated with Dey-Engley neutralizing broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth