missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMC Recombinant Mouse Monoclonal Antibody (PD01-40), Invitrogen™
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen MA550649
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive control: Rat pituitary tissue, mouse pituitary tissue. Predicted band size: 29 kDa Subcellular Location: Secreted.
This gene encodes a polypeptide hormone precursor that undergoes extensive, tissue-specific, post-translational processing via cleavage by subtilisin-like enzymes known as prohormone convertases. There are eight potential cleavage sites within the polypeptide precursor and, depending on tissue type and the available convertases, processing may yield as many as ten biologically active peptides involved in diverse cellular functions. The encoded protein is synthesized mainly in corticotroph cells of the anterior pituitary where four cleavage sites are used; adrenocorticotrophin, essential for normal steroidogenesis and the maintenance of normal adrenal weight, and lipotropin beta are the major end products. In other tissues, including the hypothalamus, placenta, and epithelium, all cleavage sites may be used, giving rise to peptides with roles in pain and energy homeostasis, melanocyte stimulation, and immune modulation. These include several distinct melanotropins, lipotropins, and endorphins that are contained within the adrenocorticotrophin and beta-lipotropin peptides. Mutations in this gene have been associated with early onset obesity, adrenal insufficiency, and red hair pigmentation. Alternatively spliced transcript variants encoding the same protein have been described.
Spécification
POMC | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P01189, P01193, P01194 | |
POMC | |
Synthetic peptide aa 1-150/267. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (Paraffin) | |
PD01-40 | |
Unconjugated | |
POMC | |
ACTH, adrenal corticotropic hormone, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte stimulating hormone, alpha-melanocyte-stimulating hormone, alphaMSH, alpha-MSH, BE, beta-endorphin, Beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, Corticotropin, Corticotropin-like intermediary peptide, corticotropin-lipotropin, Gamma-LPH, gamma-MSH, lipotr, Lipotropin beta, lipotropin gamma, LPH, Melanocyte-stimulating hormone alpha, Melanocyte-stimulating hormone beta, Melanotropin alpha, melanotropin beta, Melanotropin gamma, met-enkephalin, MSH, Npp, opiomelanocortin prepropeptide, OTTHUMP00000119991, OTTHUMP00000200964, POC, Pomc, Pomc1, Pomc-1, Pomc2, Potential peptide, Precursor of MSH, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin), proopiomelanocortin preproprotein, proopiomelanocortin, beta (endorphin, beta), pro-opiomelanocortin-alpha, proopoimelanocortin, beta (endorphin, beta) | |
Mouse | |
Protein A | |
RUO | |
18976, 24664, 5443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Suggestions de produits
Customers who viewed this item also viewed
Viewing 1-5 of 12
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
POMC Recombinant Mouse Monoclonal Antibody (PD01-40), Invitrogen™ > 100 μL; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu