Learn More
Hanna Instruments™ Portable Lux Meter
Brand:  Hanna Instruments™ HI97500
Product Code. 10542284
  • 523.00 EUR / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Specifications
Portable Lux Meter
45 mm
0.001 to 1.999
76 mm
±6% of reading ±2 digits
164 mm
180 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ Portable Lux Meter > Lux Range: 0.001 to 1.999