missing translation for 'onlineSavingsMsg'
Learn More
Potassium Chloride, Reagent Grade, Scharlab Product Code.: 11304264

Potassium Chloride, Reagent Grade, Scharlab

Product Code. 11304264
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11304264

Brand: Scharlab PO02000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, in photography, in buffer solutions, and as an electrolyte for batteries.

  • Synonym: Chloro potassium
  • Molecular Formula: KCl
  • Molar Mass: 74.56g/mol
  • CAS: 7447-40-7
  • EIN ECS No.: 231-211-8

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1413°C
Melting Point 773°C
pH 5.5–8.0 (5%, water)
Quantity 250 g
Formula Weight 74.56g/mol
Percent Purity 99.5% min (argentometric)
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride, Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.