Learn More
Potassium iodide, BAKER ANALYZED™ ACS, J.T. Baker™ Product Code.: 10000390
Potassium iodide, BAKER ANALYZED™ ACS, J.T. Baker™
Product Code. 10000390
500 g
Click to view available options
Quantity:
500 g
1 kg
Product Code. 10000390
Brand: JT Baker 0227.0500
Meets ACS Specifications
Chemical Identifiers
CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]
Specifications
Boiling Point 1330°C (1013 hPa)
Melting Point 686°C
pH 6.0 to 9.2 (5% Solution at 25°C)
Quantity 500 g
Assay Percent Range min. 99.0%
Loss on Drying max. 0.2% (at 150°C)
Density 3.13 g/cm³ (20°C)
Product Content Correction
Product Title
Potassium iodide, BAKER ANALYZED™ ACS, J.T. Baker™ >