missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Specifications
Specifications
CAS | 7778-77-0 |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | 99+% |
Molecular Formula | H2KO4P |
MDL Number | MFCD00011401 MFCD00147253 |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
SMILES | [K+].OP(O)([O-])=O |
IUPAC Name | potassium dihydrogen phosphate |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction