Learn More
Applied Biosystems™ Pouch Cap for Polymer Pouch
This is a cap for use with POP-4™, POP-6™, and POP-7™ polymer pouches.
Brand:  Applied Biosystems™ 4412619
Product Code. 16385332
Specifications
Each package contains 4 caps.
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Pouch Cap for Polymer Pouch
Room Temperature
Pouch
Sequencing
4 Pieces
Documents
One moment while we fetch your results.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP302114 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number4412619
Lot NumberCOO Certificate TypeCertificate of Analysis Date22/08/2023 Catalog Number4412619
Lot NumberP302035-001 Certificate TypeCertificate of Analysis Date20/04/2023 Catalog Number4412619
Lot NumberCOC Certificate TypeCertificate of Analysis Date26/11/2019 Catalog Number4412619
Product Title
Applied Biosystems™ Pouch Cap for Polymer Pouch >