Learn More
Hanna Instruments™ pH/EC/TDS Multiparameter Probe
Pre-amplified, multiparameter probe. Hanna Instruments™ pH/EC/TDS Multiparameter Probe measures ph, EC/TDS, and temperature within a single polypropylene body.
Brand:  Hanna Instruments™ HI1285-5
Product Code. 11692110
  • 334.00 EUR / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Description
  • Pre-amplified pH electrode
  • Amperometric EC/TDS sensor
  • Built-in temperature sensor
Order Info
Includes
Specifications
Specifications
0 to 13
Measures pH, EC/TDS, and temperature in greenhouses, hydroponics, environmental monitoring, water treatment, and boilers and cooling towers
HI-1285-5
185 mm
0°C to 50°C
Electrolyte: gel
Preamplified pH and EC Probe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ pH/EC/TDS Multiparameter Probe > Includes temperature sensor; Measures pH, EC/TDS