missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PREX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534962
This item is not returnable.
View return policy
Description
Recommended positive controls: THP-1, PREX1-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene acts as a guanine nucleotide exchange factor for the RHO family of small GTP-binding proteins (RACs). It has been shown to bind to and activate RAC1 by exchanging bound GDP for free GTP. The encoded protein, which is found mainly in the cytoplasm, is activated by phosphatidylinositol-3,4,5-trisphosphate and the beta-gamma subunits of heterotrimeric G proteins.
Specifications
PREX1 | |
Polyclonal | |
Unconjugated | |
PREX1 | |
G630042G04; KIAA1415; OTTHUMP00000031228; Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 1 protein; phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 1; phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1; PIP3 dependent Rac exchange factor 1; PREX 1; PREX1; P-Rex1; PREX-1; ptdIns(3,4,5)-dependent Rac exchanger 1; RGD1306534; SET domain containing 6; Setd6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TCU6 | |
PREX1 | |
Recombinant fragment contains a sequence corresponding to a region within amino acids 982 and 1185 of PREX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PREX1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction